Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842551.1 | XP_028698384.1 | 571 | 108 | ||
XM_028842552.1 | XP_028698385.1 | 631 | 108 | |||
GenBank mRNA | JV050744.1 | - | 643 | 108 | ||
JV635739.1 | - | 857 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |