Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116253.2 | XP_014971739.1 | 2055 | 86 | ||
GenBank mRNA | JU333872.1 | - | 409 | 86 | ||
JU333873.1 | - | 409 | 86 | |||
JU472033.1 | - | 430 | 86 | |||
JU472034.1 | - | 430 | 86 | |||
JV046684.1 | - | 560 | 86 | |||
JV046685.1 | - | 560 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |