Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYBU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265668.1 | NP_001252597.1 | 731 | 93 | ||
XM_015145878.1 | XP_015001364.1 | 1230 | 93 | |||
XM_015145880.2 | XP_015001366.1 | 1216 | 93 | |||
XM_015145881.1 | XP_015001367.1 | 1447 | 93 | |||
XM_015145882.2 | XP_015001368.1 | 1096 | 93 | |||
XM_015145884.2 | XP_015001370.1 | 1213 | 93 | |||
XM_015145885.2 | XP_015001371.1 | 1093 | 93 | |||
XM_015145889.2 | XP_015001375.1 | 809 | 93 | |||
XM_015145890.2 | XP_015001376.1 | 508 | 93 | |||
XM_028852148.1 | XP_028707981.1 | 1109 | 93 | |||
XM_028852150.1 | XP_028707983.1 | 1073 | 93 | |||
XM_028852151.1 | XP_028707984.1 | 1032 | 93 | |||
GenBank mRNA | JU335375.1 | - | 1123 | 93 | ||
JV489102.1 | - | 731 | 93 | |||
JV718449.1 | - | 913 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |