Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266028.1 | NP_001252957.1 | 429 | 161 | ||
XM_015128700.2 | XP_014984186.1 | 650 | 161 | |||
GenBank mRNA | JV064637.1 | - | 68 | 161 | ||
JV635844.1 | - | 429 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |