Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257913.1 | NP_001244842.1 | 968 | 85 | ||
XM_015137003.2 | XP_014992489.1 | 1189 | 85 | |||
XM_028846642.1 | XP_028702475.1 | 963 | 85 | |||
GenBank mRNA | JU335885.1 | - | 968 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |