Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261085.1 | NP_001248014.1 | 969 | 69 | ||
GenBank mRNA | JU477194.1 | - | 1020 | 69 | ||
JV048246.1 | - | 969 | 69 | |||
JV481759.1 | - | 510 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |