Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266713.1 | NP_001253642.1 | 4113 | 70 | ||
XM_015118795.2 | XP_014974281.1 | 4321 | 70 | |||
XM_015118805.2 | XP_014974291.1 | 4033 | 70 | |||
XM_015118811.2 | XP_014974297.1 | 4240 | 70 | |||
XM_028843550.1 | XP_028699383.1 | 3814 | 70 | |||
GenBank mRNA | JU322212.1 | - | 4212 | 70 | ||
JV489114.1 | - | 4113 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |