Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124715.2 | XP_014980201.1 | 1667 | 124 | ||
GenBank mRNA | JU327901.1 | - | 208 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Macaca mulatta |