Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265847.1 | NP_001252776.1 | 449 | 82 | ||
XM_015132568.2 | XP_014988054.1 | 317 | 82 | |||
XM_015132569.1 | XP_014988055.1 | 548 | 82 | |||
XM_015132571.1 | XP_014988057.1 | 552 | 82 | |||
XM_015132572.2 | XP_014988058.1 | 520 | 82 | |||
XM_028843283.1 | XP_028699116.1 | 608 | 82 | |||
GenBank mRNA | JU319782.1 | - | 467 | 82 | ||
JU473073.1 | - | 450 | 82 | |||
JV045570.1 | - | 449 | 82 | |||
JV529474.1 | - | 404 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |