Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260706.1 | NP_001247635.1 | 300 | 78 | ||
XM_015118616.2 | XP_014974102.1 | 541 | 78 | |||
XM_015118617.2 | XP_014974103.1 | 430 | 78 | |||
XM_015118618.2 | XP_014974104.1 | 432 | 78 | |||
XM_015118619.2 | XP_014974105.1 | 468 | 78 | |||
XM_028835463.1 | XP_028691296.1 | 1451 | 78 | |||
XM_028835464.1 | XP_028691297.1 | 541 | 78 | |||
GenBank mRNA | JU322751.1 | - | 345 | 78 | ||
JU475807.1 | - | 263 | 78 | |||
JV047232.1 | - | 300 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |