Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114114.1 | NP_001107586.1 | 149 | 73 | ||
GenBank mRNA | EU338447.1 | - | 149 | 73 |