Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080762.1 | NP_001074231.1 | 354 | 68 | ||
XM_015127870.2 | XP_014983356.1 | 614 | 68 | |||
XM_015127871.2 | XP_014983357.1 | 602 | 68 | |||
XM_015127873.2 | XP_014983359.1 | 612 | 68 | |||
XM_015127874.2 | XP_014983360.1 | 594 | 68 | |||
XM_015127875.2 | XP_014983361.1 | 607 | 68 | |||
XM_028841674.1 | XP_028697507.1 | 624 | 68 | |||
XM_028841676.1 | XP_028697509.1 | 606 | 68 | |||
XM_028841677.1 | XP_028697510.1 | 516 | 68 | |||
GenBank mRNA | JU337132.1 | - | 522 | 68 | ||
JU337133.1 | - | 510 | 68 | |||
JU477871.1 | - | 509 | 68 | |||
JV048720.1 | - | 529 | 68 | |||
JV048721.1 | - | 517 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |