Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194747.1 | NP_001181676.1 | 75 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |