Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194889.2 | NP_001181818.1 | 468 | 87 | ||
GenBank mRNA | JU320925.1 | - | 467 | 87 | ||
JU472676.1 | - | 436 | 87 | |||
JV669643.1 | - | 428 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |