Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194893.2 | NP_001181822.1 | 725 | 102 | ||
XM_002804966.3 | XP_002805012.1 | 741 | 102 | |||
GenBank mRNA | JU337017.1 | - | 503 | 102 | ||
JU337018.1 | - | 524 | 102 | |||
JU472496.1 | - | 549 | 102 | |||
JU472497.1 | - | 528 | 102 | |||
JV525112.1 | - | 401 | 102 | |||
JV667681.1 | - | 534 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |