Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258156.1 | NP_001245085.1 | 1821 | 85 | ||
GenBank mRNA | JU327547.1 | - | 1303 | 85 | ||
JU473218.1 | - | 1821 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |