Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265666.1 | NP_001252595.1 | 985 | 80 | ||
XM_015121917.2 | XP_014977403.1 | 1919 | 80 | |||
XM_015121918.2 | XP_014977404.1 | 1727 | 80 | |||
XM_015121919.2 | XP_014977405.1 | 1831 | 80 | |||
XM_015121920.2 | XP_014977406.1 | 1752 | 80 | |||
XM_015121921.2 | XP_014977407.1 | 1795 | 80 | |||
XM_028837715.1 | XP_028693548.1 | 2012 | 80 | |||
XM_028837716.1 | XP_028693549.1 | 1976 | 80 | |||
XM_028837717.1 | XP_028693550.1 | 1768 | 80 | |||
XM_028837718.1 | XP_028693551.1 | 2009 | 80 | |||
GenBank mRNA | JV566742.1 | - | 1692 | 80 | ||
JV635691.1 | - | 985 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |