Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265877.1 | NP_001252806.1 | 189 | 93 | ||
GenBank mRNA | JV599139.1 | - | 245 | 93 | ||
JV667510.1 | - | 189 | 93 |