Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266683.1 | NP_001253612.1 | 355 | 73 | ||
XM_015135190.2 | XP_014990676.1 | 851 | 73 | |||
XM_015135191.2 | XP_014990677.1 | 403 | 73 | |||
GenBank mRNA | JU473731.1 | - | 299 | 73 | ||
JV047825.1 | - | 350 | 73 | |||
JV636301.1 | - | 355 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |