Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017024.2 | NP_058720.2 | 1 - 2 | 166 | 70 | |
GenBank mRNA | BC091155.1 | - | 1 - 2 | 166 | 7370790 | 70 |
FQ209624.1 | - | 1 - 2 | 181 | 70 | ||
FQ218245.1 | - | 1 - 2 | 181 | 70 | ||
FQ218883.1 | - | 1 - 2 | 184 | 70 | ||
U62803.1 | - | 1 - 2 | 169 | 70 | ||
X54096.1 | - | 1 - 2 | 180 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |