Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022529.1 | NP_071974.1 | 3 - 4 | 236 | 104 | |
GenBank mRNA | BC059936.1 | - | 3 - 4 | 242 | 104 | |
U62635.1 | - | 3 - 4 | 236 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |