Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031667.2 | NP_113855.2 | 1 - 2 | 235 | 73 | |
XM_006232748.1 | XP_006232810.1 | 1 - 2 | 227 | 73 | ||
GenBank mRNA | AF000423.1 | - | 1 - 2 | 276 | 73 | |
BC094518.1 | - | 1 - 2 | 235 | 7315033 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |