Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053955.1 | NP_446407.1 | 7 - 8 | 906 | 89 | |
GenBank mRNA | BC088121.1 | - | 7 - 8 | 937 | 7323247 | 89 |
Y17328.1 | - | 7 - 8 | 906 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |