Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057098.1 | NP_476439.1 | 4 - 5 | 411 | 59 | |
XM_006235741.1 | XP_006235803.1 | 4 - 5 | 488 | 59 | ||
GenBank mRNA | D12927.1 | - | 4 - 5 | 411 | 59 | |
FQ213752.1 | - | 4 - 5 | 389 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons |