Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1952 | 93 | |
GenBank mRNA | AF032669.1 | - | 1 | 1952 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |