Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145673.2 | NP_663706.2 | 2 - 3 | 230 | 82 | |
XM_006248915.2 | XP_006248977.1 | 2 - 3 | 3471 | 82 | ||
XM_006248917.2 | XP_006248979.1 | 2 - 3 | 104 | 82 | ||
GenBank mRNA | AF461686.1 | - | 2 - 3 | 145 | 82 | |
BC070906.1 | - | 2 - 3 | 230 | 7103075 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |