Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175760.2 | NP_786936.1 | 201 | 64 | ||
GenBank mRNA | M33936.1 | - | 201 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 P450 Rattus norvegicus Rat Androgen Panel Inventoried Probe spans exons |