Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031634.1 | NP_113822.1 | 7 - 8 | 1957 | 73 | |
GenBank mRNA | AF143410.1 | - | 7 - 8 | 1957 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |