Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022606.1 | NP_072128.1 | 2 - 3 | 165 | 88 | |
GenBank mRNA | AF095927.1 | - | 2 - 3 | 165 | 88 | |
BC062010.1 | - | 2 - 3 | 167 | 5621497 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |