Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138523.2 | NP_612532.1 | 6 - 7 | 676 | 95 | |
GenBank mRNA | AF244920.1 | - | 6 - 7 | 671 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |