Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139188.3 | NP_631927.1 | 2 - 3 | 116 | 72 | |
XM_008767358.1 | XP_008765580.1 | 3 - 4 | 724 | 72 | ||
GenBank mRNA | AY062254.1 | - | 2 - 3 | 144 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |