Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017223.2 | NP_058919.1 | 8 - 9 | 1936 | 73 | |
XM_008771351.1 | XP_008769573.1 | 8 - 9 | 2943 | 73 | ||
XM_008771352.1 | XP_008769574.1 | 8 - 9 | 2583 | 73 | ||
GenBank mRNA | BC070908.1 | - | 8 - 9 | 1936 | 7103640 | 73 |
L19931.1 | - | 7 - 8 | 1792 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |