Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017014.1 | NP_058710.1 | 5 - 6 | 397 | 72 | |
GenBank mRNA | BC063172.1 | - | 5 - 6 | 437 | 6920688 | 72 |
FJ179401.1 | - | 5 - 6 | 361 | 72 | ||
FQ211067.1 | - | 5 - 6 | 396 | 72 | ||
FQ212742.1 | - | 5 - 6 | 390 | 72 | ||
FQ213204.1 | - | 5 - 6 | 390 | 72 | ||
FQ213696.1 | - | 5 - 6 | 584 | 72 | ||
FQ214055.1 | - | 5 - 6 | 496 | 72 | ||
FQ217092.1 | - | 5 - 6 | 463 | 72 | ||
FQ218789.1 | - | 5 - 6 | 396 | 72 | ||
FQ218795.1 | - | 5 - 6 | 433 | 72 | ||
FQ229836.1 | - | 5 - 6 | 444 | 72 | ||
FQ229941.1 | - | 5 - 6 | 424 | 72 | ||
J02810.1 | - | 5 - 6 | 397 | 72 | ||
M11719.1 | - | 5 - 6 | 398 | 72 | ||
M28241.1 | - | 5 - 6 | 457 | 72 | ||
X04229.1 | - | 5 - 6 | 397 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |