Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080766.2 | NP_542944.1 | 4 - 5 | 557 | 82 | |
XM_006233059.2 | XP_006233121.1 | 4 - 5 | 601 | 82 | ||
GenBank mRNA | BC098659.1 | - | 4 - 5 | 557 | 7368145 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |