Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005875.1 | NP_001005875.1 | 7 - 8 | 852 | 72 | |
GenBank mRNA | BC083758.1 | - | 7 - 8 | 852 | 7192849 | 72 |
FQ218292.1 | - | 1 - 2 | 79 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |