Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181367.2 | NP_852032.1 | 3 - 4 | 274 | 72 | |
XM_006249914.2 | XP_006249976.1 | 1 - 2 | 766 | 72 | ||
XM_006249915.2 | XP_006249977.1 | 1 - 2 | 771 | 72 | ||
XM_006249916.2 | XP_006249978.1 | 4 - 5 | 836 | 72 | ||
XM_006249917.1 | XP_006249979.1 | 1 - 2 | 127 | 72 | ||
XM_006249918.2 | XP_006249980.1 | 1 - 2 | 761 | 72 | ||
XM_006249919.2 | XP_006249981.1 | 4 - 5 | 836 | 72 | ||
XM_008769545.1 | XP_008767767.1 | 3 - 4 | 362 | 72 | ||
XM_008769546.1 | XP_008767768.1 | 1 - 2 | 761 | 72 | ||
GenBank mRNA | AF527619.1 | - | 1 - 2 | 82 | 72 | |
AY273890.1 | - | 3 - 4 | 274 | 72 | ||
AY724502.1 | - | 3 - 4 | 274 | 72 | ||
BC128722.1 | - | 4 - 5 | 350 | 8362986 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |