Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108470.1 | NP_001101940.1 | 1 - 2 | 873 | 62 | |
XM_006228116.2 | XP_006228178.1 | 2 - 3 | 760 | 62 | ||
XM_008758767.1 | XP_008756989.1 | 2 - 3 | 1021 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |