Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105902.1 | NP_001099372.1 | 2 - 3 | 302 | 81 | |
XM_008768945.1 | XP_008767167.1 | 2 - 3 | 478 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |