Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183329.3 | NP_899158.3 | 4 - 5 | 511 | 78 | |
GenBank mRNA | AY026354.1 | - | 4 - 5 | 459 | 78 | |
AY542868.1 | - | 4 - 5 | 508 | 78 | ||
BC091105.1 | - | 3 - 4 | 1295 | 7381969 | 78 | |
CB322787.1 | - | 1 - 2 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |