Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108107.2 | NP_001101577.2 | 1 - 2 | 309 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |