Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079939.1 | NP_001073408.1 | 11 - 12 | 1750 | 84 | |
GenBank mRNA | BC088333.1 | - | 1 - 2 | 163 | 84 | |
BC129102.1 | - | 11 - 12 | 1750 | 7099869 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |