Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ribc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007715.1 | NP_001007716.1 | 7 - 8 | 1225 | 78 | |
XM_006256799.2 | XP_006256861.1 | 6 - 7 | 1412 | 78 | ||
XM_006256800.2 | XP_006256862.1 | 7 - 8 | 1365 | 78 | ||
XM_006256801.2 | XP_006256863.1 | 7 - 8 | 1376 | 78 | ||
XM_006256802.2 | XP_006256864.1 | 7 - 8 | 1280 | 78 | ||
XM_006256803.2 | XP_006256865.1 | 7 - 8 | 1283 | 78 | ||
XM_006256804.2 | XP_006256866.1 | 7 - 8 | 1265 | 78 | ||
XM_006256805.2 | XP_006256867.1 | 7 - 8 | 1261 | 78 | ||
GenBank mRNA | BC078998.1 | - | 7 - 8 | 1225 | 7112763 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |