Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250950.2 | XP_006251012.1 | 1 - 2 | 1982 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |