Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100510.1 | NP_001093980.1 | 10 - 11 | 1052 | 66 | |
GenBank mRNA | BC087140.1 | - | 9 - 10 | 1117 | 66 | |
BC158592.1 | - | 10 - 11 | 1052 | 8362956 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |