Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 9 - 10 | 971 | 88 | |
GenBank mRNA | BC158690.1 | - | 9 - 10 | 971 | 8376316 | 88 |
BF414694.1 | - | 1 - 2 | 88 | |||
BI282626.1 | - | 1 - 2 | 88 | |||
FQ216026.1 | - | 9 - 10 | 966 | 88 | ||
FQ216320.1 | - | 9 - 10 | 947 | 88 | ||
FQ230458.1 | - | 10 - 11 | 816 | 88 | ||
FQ233169.1 | - | 9 - 10 | 948 | 88 | ||
FQ234620.1 | - | 9 - 10 | 969 | 88 | ||
FQ234801.1 | - | 9 - 10 | 947 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |