Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053409.2 | NP_445861.1 | 8 - 9 | 1855 | 85 | |
GenBank mRNA | AF217964.1 | - | 8 - 9 | 1991 | 85 | |
AF274043.1 | - | 6 - 7 | 1194 | 85 | ||
AJ133038.1 | - | 6 - 7 | 1318 | 85 | ||
BC081756.1 | - | 8 - 9 | 1855 | 7134252 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |