Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104643.1 | NP_001098113.1 | 4 - 5 | 326 | 66 | |
GenBank mRNA | EU074986.1 | - | 4 - 5 | 326 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |