Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ229819.1 | - | 147 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |