Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019377.1 | NP_062250.1 | 3 - 4 | 570 | 116 | |
GenBank mRNA | BC076502.1 | - | 3 - 4 | 564 | 7100393 | 116 |
D17446.1 | - | 3 - 4 | 565 | 116 | ||
FQ214109.1 | - | 3 - 4 | 606 | 116 | ||
FQ229978.1 | - | 3 - 4 | 575 | 116 | ||
S55223.1 | - | 3 - 4 | 565 | 116 | ||
S83440.1 | - | 3 - 4 | 570 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |