Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053708.2 | NP_446160.1 | 1 - 2 | 561 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |